Anti-BSA Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-0292R
BOSSBS-0292REA 8800 NOK
BOSSBS-0292R
Anti-BSA Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR