Anti-BSA Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-0292R-A680
BOSSBS-0292R-A680EA 10525 NOK
BOSSBS-0292R-A680
Anti-BSA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Serum albumin, the main protein of plasma, has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood. Major zinc transporter in plasma, typically binds about 8% of all plasma zinc.

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Goat, Others, Cow
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR