Anti-BSA Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-0292R-A750
BOSSBS-0292R-A750EA 10525 NOK
BOSSBS-0292R-A750
Anti-BSA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Serum albumin, the main protein of plasma, has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood. Major zinc transporter in plasma, typically binds about 8% of all plasma zinc.

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Goat, Others, Cow
Bestill nå

SPESIFIKASJONER

Antigen-navn BSA
Environmentally Preferable
Antigen synonymer Serum albumin|BSA|ALB
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Goat,Human,Bovine,Rat,Mouse
Vert Rabbit
Gen-ID 280717
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Immunogen BSA
Skjema Liquid
Molekylvekt 69 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR