Anti-ITGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-0486R-A350
BOSSBS-0486R-A350EA 10550 NOK
BOSSBS-0486R-A350
Anti-ITGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Integrins alpha-1/beta-1, alpha-2/beta-1, alpha-10/beta-1 and alpha-11/beta-1 are receptors for collagen. Integrins alpha-1/beta-1 and alpha-2/beta-2 recognize the proline-hydroxylated sequence G-F-P-G-E-R in collagen. Integrins alpha-2/beta-1, alpha-3/beta-1, alpha-4/beta-1, alpha-5/beta-1, alpha-8/beta-1, alpha-10/beta-1, alpha-11/beta-1 and alpha-V/beta-1 are receptors for fibronectin. Alpha-4/beta-1 recognizes one or more domains within the alternatively spliced CS-1 and CS-5 regions of fibronectin. Integrin alpha-5/beta-1 is a receptor for fibrinogen. Integrin alpha-1/beta-1, alpha-2/More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol ITGB1
Antigen-navn Integrin Beta 1
Environmentally Preferable
Antigen synonymer FNRB|integrin VLA-4 beta subunit|VLA-BETA|VLAB|VLA-4 subunit beta|CD29|beta polypeptide|MDF2|MSK12|GPIIA|Fibronectin receptor subunit beta|very late activation protein
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 3688
Isotype IgG
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen 10-60/798
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR