Anti-Integrin beta 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-0486R-A680
BOSSBS-0486R-A680EA 10525 NOK
BOSSBS-0486R-A680
Anti-Integrin beta 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Integrins alpha-1/beta-1, alpha-2/beta-1, alpha-10/beta-1 and alpha-11/beta-1 are receptors for collagen. Integrins alpha-1/beta-1 and alpha-2/beta-2 recognise the proline-hydroxylated sequence G-F-P-G-E-R in collagen. Integrins alpha-2/beta-1, alpha-3/beta-1, alpha-4/beta-1, alpha-5/beta-1, alpha-8/beta-1, alpha-10/beta-1, alpha-11/beta-1 and alpha-V/beta-1 are receptors for fibronectin. Alpha-4/beta-1 recognises one or more domains within the alternatively spliced CS-1 and CS-5 regions of fibronectin. Integrin alpha-5/beta-1 is a receptor for fibrinogen. Integrin alpha-1/beta-1, alpha-2/More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Integrin beta 1
Antigen synonymer FNRB|Glycoprotein IIa|VLA-BETA|VLAB|VLA-4 subunit beta|CD29|MDF2|Integrin beta-1|MSK12|GPIIA|Fibronectin receptor subunit beta|ITGB1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet Rabbit,Sheep,Human,Rat,Mouse
Vert Rabbit
Gen-ID 3688
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Modifikasjon Unmodified
Epitop 25-100/798
Immunogen KLH conjugated synthetic peptide derived from human Integrin beta 1
Skjema Liquid
Molekylvekt 88 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR