Anti-ITGB1 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-0486R-CY7
BOSSBS-0486R-CY7EA 10650 NOK
BOSSBS-0486R-CY7
Anti-ITGB1 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Integrins alpha-1/beta-1, alpha-2/beta-1, alpha-10/beta-1 and alpha-11/beta-1 are receptors for collagen. Integrins alpha-1/beta-1 and alpha-2/beta-2 recognize the proline-hydroxylated sequence G-F-P-G-E-R in collagen. Integrins alpha-2/beta-1, alpha-3/beta-1, alpha-4/beta-1, alpha-5/beta-1, alpha-8/beta-1, alpha-10/beta-1, alpha-11/beta-1 and alpha-V/beta-1 are receptors for fibronectin. Alpha-4/beta-1 recognizes one or more domains within the alternatively spliced CS-1 and CS-5 regions of fibronectin. Integrin alpha-5/beta-1 is a receptor for fibrinogen. Integrin alpha-1/beta-1, alpha-2/More Product Information