Anti-ABCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-0657R-A647
BOSSBS-0657R-A647EA 10550 NOK
BOSSBS-0657R-A647
Anti-ABCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
Mediates export of organic anions and drugs from the cytoplasm. Mediates ATP-dependent transport of glutathione and glutathione conjugates, leukotriene C4, estradiol-17-beta-o-glucuronide, methotrexate, antiviral drugs and other xenobiotics. Confers resistance to anticancer drugs. Hydrolyzes ATP with low efficiency.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR