Anti-MRP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-0657R-A750
BOSSBS-0657R-A750EA 10525 NOK
BOSSBS-0657R-A750
Anti-MRP1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Mediates export of organic anions and drugs from the cytoplasm. Mediates ATP-dependent transport of glutathione and glutathione conjugates, leukotriene C4, estradiol-17-beta-o-glucuronide, methotrexate, antiviral drugs and other xenobiotics. Confers resistance to anticancer drugs. Hydrolyses ATP with low efficiency.

Type: Primary
Antigen: MRP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/1531
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR