Anti-HHV8 ORF50 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-0746R-CY5
BOSSBS-0746R-CY5EA 10275 NOK
BOSSBS-0746R-CY5
Anti-HHV8 ORF50 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
HHV8 has been found to be associated with three different diseases observed in AIDS patients; kaposi's sarcoma, primary effusion lymphoma (which is a rare type of non-Hodgkin lymphoma affecting the body cavities) and multicentric Castleman's disease. To date there is much evidence to support a direct role for HHV8 in kaposi's sarcoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HHV8 ORF50
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR