Anti-HHV8 ORF50 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-0859R-CY3
BOSSBS-0859R-CY3EA 9500 NOK
BOSSBS-0859R-CY3
Anti-HHV8 ORF50 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
HHV8 has been found to be associated with three different diseases observed in AIDS patients; kaposi's sarcoma, primary effusion lymphoma (which is a rare type of non-Hodgkin lymphoma affecting the body cavities) and multicentric Castleman's disease. To date there is much evidence to support a direct role for HHV8 in kaposi's sarcoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HHV8 ORF50
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestill nå

SPESIFIKASJONER

Antigensymbol HHV8 ORF50
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy3®
Reaktivitet Virus
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR