Anti-2,4-D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Bioss
BS-0861R-HRP
BOSSBS-0861R-HRPEA 10550 NOK
BOSSBS-0861R-HRP
Anti-2,4-D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
2,4-Dichlorophenoxyacetic acid (2,4-D) is a common systemic herbicide used in the control of broadleaf weeds. It is the most widely used herbicide in the world, and the third most commonly used in North America. [1] 2,4-D is also an important synthetic auxin, often used in laboratories for plant research and as a supplement in plant cell culture media such as MS medium. 2,4-D is a synthetic auxin, which is a class of plant growth regulators. It is absorbed through the leaves and is translocated to the meristems of the plant. Uncontrolled, unsustainable growth ensues causing stem curl-over,More Product Information