Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-10141R-A555
BOSSBS-10141R-A555EA 10450 NOK
BOSSBS-10141R-A555
Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR