Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-10141R-A750
BOSSBS-10141R-A750EA 10425 NOK
BOSSBS-10141R-A750
Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-550/638
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR