Anti-CHI3L1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-10215R-A680
BOSSBS-10215R-A680EA 10250 NOK
BOSSBS-10215R-A680
Anti-CHI3L1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Carbohydrate-binding lectin with a preference for chitin. Has no chitinase activity. May play a role in tissue remodeling and in the capacity of cells to respond to and cope with changes in their environment. Plays a role in T-helper cell type 2 (Th2) inflammatory response and IL-13-induced inflammation, regulating allergen sensitization, inflammatory cell apoptosis, dendritic cell accumulation and M2 macrophage differentiation. Facilitates invasion of pathogenic enteric bacteria into colonic mucosa and lymphoid organs. Mediates activation of AKT1 Signalling pathway and subsequent IL8More Product Information