Anti-TREM1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-10306R
BOSSBS-10306REA 8500 NOK
BOSSBS-10306R
Anti-TREM1 Rabbit Polyclonal Antibody
Antistoffer
Stimulates neutrophil and monocyte-mediated inflammatory responses. Triggers release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. Amplifier of inflammatory responses that are triggered by bacterial and fungal infections and is a crucial mediator of septic shock.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TREM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol TREM1
Antigen-navn Triggering receptor expressed on myeloid cells 1
Environmentally Preferable
Antigen synonymer Trem1|CD354|TREM-1|Triggering receptor expressed on myeloid cells 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 54210
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunogen 150-200/234
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR