Anti-TREM1 Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-10306R-CY3
BOSSBS-10306R-CY3EA 10550 NOK
BOSSBS-10306R-CY3
Anti-TREM1 Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Stimulates neutrophil and monocyte-mediated inflammatory responses. Triggers release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. Amplifier of inflammatory responses that are triggered by bacterial and fungal infections and is a crucial mediator of septic shock.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TREM1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR