Anti-Galectin-3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Leverandør: Bioss
BS-10377R-A750
BOSSBS-10377R-A750EA 10425 NOK
BOSSBS-10377R-A750
Anti-Galectin-3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 750)
Antistoffer
This gene encodes a member of the galectin family of carbohydrate binding proteins. Members of this protein family have an affinity for beta-galactosides. The encoded protein is characterised by an N-terminal proline-rich tandem repeat domain and a single C-terminal carbohydrate recognition domain. This protein can self-associate through the N-terminal domain allowing it to bind to multivalent saccharide ligands. This protein localises to the extracellular matrix, the cytoplasm and the nucleus. This protein plays a role in numerous cellular functions including apoptosis, innate immunity,More Product Information