Anti-PCDHB12 Rabbit Polyclonal Antibody
Leverandør: Bioss
BS-11153R
BOSSBS-11153REA 8500 NOK
BOSSBS-11153R
Anti-PCDHB12 Rabbit Polyclonal Antibody
Antistoffer
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHB12 (Protocadherin beta 12) is a 795 amino acid protein that is one of 16 proteins in the protocadherin beta cluster. Unlike the alpha and gamma gene clusters whose genes are spliced to downstream constant region exons during transcription, members of the beta cluster (such as PCDHB12) do notMore Product Information