Anti-PCDHB12 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-11153R-A680
BOSSBS-11153R-A680EA 10425 NOK
BOSSBS-11153R-A680
Anti-PCDHB12 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHB12 (Protocadherin beta 12) is a 795 amino acid protein that is one of 16 proteins in the protocadherin beta cluster. Unlike the alpha and gamma gene clusters whose genes are spliced to downstream constant region exons during transcription, members of the beta cluster (such as PCDHB12) do notMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn PCDHB12
Environmentally Preferable
Antigen synonymer PCDHB12|PCDH beta12|PCDBC_HUMAN|PCDHB 12
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet Human
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 121-220/795
Immunogen KLH conjugated synthetic peptide derived from human PCDHB12
Skjema Liquid
Molekylvekt 84 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR