Anti-PAF Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-1119R-A680
BOSSBS-1119R-A680EA 10425 NOK
BOSSBS-1119R-A680
Anti-PAF Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Azelaoyl PAF is a potent PPAR_ agonist which competes for the thiazolidinedione binding site.

Type: Primary
Antigen: PAF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Others
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR