Anti-PAF Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-1119R-CY5.5
BOSSBS-1119R-CY5.5EA 10550 NOK
BOSSBS-1119R-CY5.5
Anti-PAF Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
Azelaoyl PAF is a potent PPARγ agonist which competes for the thiazolidinedione binding site.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAF
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestill nå

SPESIFIKASJONER

Antigensymbol PAF
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Multiple
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR