Anti-PAF Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-1119R-CY7
BOSSBS-1119R-CY7EA 10550 NOK
BOSSBS-1119R-CY7
Anti-PAF Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Azelaoyl PAF is a potent PPARγ agonist which competes for the thiazolidinedione binding site.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAF
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR