Anti-HSD17B4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-11296R-A750
BOSSBS-11296R-A750EA 10250 NOK
BOSSBS-11296R-A750
Anti-HSD17B4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
17Beta-HSD4 (17Beta-hydroxysteroid dehydrogenase type 4) is also known as peroxisomal multifunctional enzyme/protein 2 (MFE-2/MFP-2), D-bifunctional enzyme or 17-Beta Estradiol dehydrogenase type IV. It belongs to the 17Beta-HSD family of proteins that regulate the availability of steroids within various tissues throughout the body. 17Beta-HSD4 inactivates Estradiol through its oxidative activity but it is primarily involved in peroxisomal fatty acid and cholesterol Beta-oxidation. It has a multi-domain structure: the dehydrogenase domain is fused to a hydratase and a lipid transfer domain. 1More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn HSD17B4
Antigen synonymer MFE-2|17 beta hydroxysteroid dehydrogenase 4|D bunctional protein peroxisomal|Peroxisomal multunctional protein 2|Hsd17b4|3 alpha 7 alpha12 alpha trihydroxy 5 beta cholest 24 enoyl CoA hydratase antibody 3-alpha|MFE 2|EDH17B4|17beta estradiol dehydrogenase type IV|17-beta-hydroxysteroid dehydrogenase 4|7-alpha|D-bunctional protein|Peroxisomal multunctional enzyme type 2|17 beta HSD IV|D 3 hydroxyacyl CoA dehydratase|17 beta HSD 4|17-beta-HSD 4|D bunctional protein|DHB4_HUMAN|Short chain dehydrogenase/reductase family 8C member 1|12-alpha-trihydroxy-5-beta-cholest-24-enoyl-CoA hydratase|DBP|SDR8C1|Beta keto reductase|Beta hydroxyacyl dehydrogenase|MPF-2|Enoyl-CoA hydratase 2|Multunctional protein 2|hydroxysteroid 17-beta dehydrogenase 4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Zebrafish,Sheep,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 521-620/736
Immunogen KLH conjugated synthetic peptide derived from human HSD17B4 Enoyl-CoA hydratase 2
Skjema Liquid
Molekylvekt 47/80 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR