Anti-HSD17B4 Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-11296R-CY5
BOSSBS-11296R-CY5EA 10350 NOK
BOSSBS-11296R-CY5
Anti-HSD17B4 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
17Beta-HSD4 (17Beta-hydroxysteroid dehydrogenase type 4) is also known as peroxisomal multifunctional enzyme/protein 2 (MFE-2/MFP-2), D-bifunctional enzyme or 17-Beta Estradiol dehydrogenase type IV. It belongs to the 17Beta-HSD family of proteins that regulate the availability of steroids within various tissues throughout the body. 17Beta-HSD4 inactivates Estradiol through its oxidative activity but it is primarily involved in peroxisomal fatty acid and cholesterol Beta-oxidation. It has a multi-domain structure: the dehydrogenase domain is fused to a hydratase and a lipid transfer domain. 1More Product Information