Anti-BCAS1/NABC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-11462R-A680
BOSSBS-11462R-A680EA 10425 NOK
BOSSBS-11462R-A680
Anti-BCAS1/NABC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
NaBC1 is a protein found amplified in most breast carcinoma forms. It is expressed primarily as a cytoplasmic, detergent-stable homodimer that has a tendency to interact with DYNLL1 (PIN) and DYNLL2. Breast tumour lines that exhibit 20q13.2 gene amplification express much higher levels of the protein as compared to the levels found in other breast cancer lines that do not overexpress the NaBC1 mRNA. However, this upregulation does not affect growth rate or anchoring abilities of a cell, indicating the oncogenic properties of NaBC1 differ from that of other oncogenes.

Type: Primary
Antigen:More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn BCAS1/NABC1
Antigen synonymer Breast carcinoma amplied sequence 1|BCAS 1|Amplied and overexpressed in breast cancer|Breast carcinoma-amplied sequence 1|AIBC 1|BCAS1_HUMAN|AIBC1|BCAS1|Novel amplied in breast cancer 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 542-584/584
Immunogen KLH conjugated synthetic peptide derived from human BCAS1
Skjema Liquid
Molekylvekt 62 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR