Anti-LRRTM3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-11571R-A350
BOSSBS-11571R-A350EA 10450 NOK
BOSSBS-11571R-A350
Anti-LRRTM3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic Alpha/Beta horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. The LRRTM protein family plays a role in the regulation of various cellular events during nervous system development. Localizing predominantly to the nervous system, LRRTM family members are known to exhibit synaptogenic activity. LRRTM3 (leucine richMore Product Information