Anti-AGEs Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Leverandør: Bioss
BS-1158R-A647
BOSSBS-1158R-A647EA 10550 NOK
BOSSBS-1158R-A647
Anti-AGEs Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
Serum albumin, the main protein of plasma, has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood. Major zinc transporter in plasma, typically binds about 8% of all plasma zinc.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGEs
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR