Anti-AGEs Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-1158R-FITC
BOSSBS-1158R-FITCEA 10650 NOK
BOSSBS-1158R-FITC
Anti-AGEs Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Serum albumin, the main protein of plasma, has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood. Major zinc transporter in plasma, typically binds about 8% of all plasma zinc.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGEs
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol AGEs
Environmentally Preferable
Antigen-navn Advanced glycation end products
Antistofftype Primary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 280717
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR