Anti-RGS21 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-11620R-A350
BOSSBS-11620R-A350EA 10450 NOK
BOSSBS-11620R-A350
Anti-RGS21 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
The regulators of G protein signaling (RGS) proteins inhibit heterotrimeric G protein signaling. RGS proteins work by functioning as GTPase-activators (which increase the GTPase activity of G protein a-subunits) thereby driving G proteins into their inactive GDP-bound form. RGS21 (regulator of G-protein signaling 21) is a 152 amino acid protein that is ubiquitously expressed and contains one RGS domain. The gene encoding RGS21 maps to human chromosome 1q31.2 and mouse chromosome 1 F. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the humanMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol RGS21
Antigen-navn Regulator of G-protein Signaling 21
Environmentally Preferable
Antigen synonymer Regulator of G-protein signaling 21|RGS21|RGS21_HUMAN.
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR