Anti-NPEPPS Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-11684R-A555
BOSSBS-11684R-A555EA 10450 NOK
BOSSBS-11684R-A555
Anti-NPEPPS Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Puromycin-sensitive aminopeptidase is a 100kDa zinc metallopeptidase which degrades neuropeptides by removing amino acid residues from the amino-terminus. The protein is the most abundant aminopeptidase in the brain, however it is not exclusive to that organ. It is localized primarily in the cytoplasm, and plays a role in the metabolism of neuropeptides in nerve terminals and synaptic clefts. The human PSA gene maps to chromosome 17q 2-32.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPEPPS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR