Anti-NPEPPS/PSA Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-11684R-A750
BOSSBS-11684R-A750EA 10425 NOK
BOSSBS-11684R-A750
Anti-NPEPPS/PSA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Puromycin-sensitive aminopeptidase is a 100 kDa zinc metallopeptidase which degrades neuropeptides by removing amino acid residues from the amino-terminus. The protein is the most abundant aminopeptidase in the brain, however it is not exclusive to that organ. It is localised primarily in the cytoplasm, and plays a role in the metabolism of neuropeptides in nerve terminals and synaptic clefts. The human PSA gene maps to chromosome 17q 2-32.

Type: Primary
Antigen: NPEPPS/PSA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/919
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR