Anti-NPEPPS Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-11684R-CY7
BOSSBS-11684R-CY7EA 10550 NOK
BOSSBS-11684R-CY7
Anti-NPEPPS Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Puromycin-sensitive aminopeptidase is a 100kDa zinc metallopeptidase which degrades neuropeptides by removing amino acid residues from the amino-terminus. The protein is the most abundant aminopeptidase in the brain, however it is not exclusive to that organ. It is localized primarily in the cytoplasm, and plays a role in the metabolism of neuropeptides in nerve terminals and synaptic clefts. The human PSA gene maps to chromosome 17q 2-32.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPEPPS
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR