Anti-NPEPPS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Bioss
BS-11684R-HRP
BOSSBS-11684R-HRPEA 10550 NOK
BOSSBS-11684R-HRP
Anti-NPEPPS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Puromycin-sensitive aminopeptidase is a 100kDa zinc metallopeptidase which degrades neuropeptides by removing amino acid residues from the amino-terminus. The protein is the most abundant aminopeptidase in the brain, however it is not exclusive to that organ. It is localized primarily in the cytoplasm, and plays a role in the metabolism of neuropeptides in nerve terminals and synaptic clefts. The human PSA gene maps to chromosome 17q 2-32.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NPEPPS
Clonality: Polyclonal
Clone:
Conjugation: HRP (More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR