Anti-HAAO Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-11695R-A488
BOSSBS-11695R-A488EA 10450 NOK
BOSSBS-11695R-A488
Anti-HAAO Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
The 3-HAO family of proteins consists of intramolecular dioxygenases that contain non-heme ferrous iron. HAAO (3-hydroxyanthranilate 3,4-dioxygenase), also known as HAO or 3-HAO, is a 286 amino acid monomeric cytoplasmic protein that belongs to the 3-HAO family. Utilizing Fe2+ ions as a cofactor, HAAO participates in the formation of quinolinic acid (QUIN), and excitotoxin, by catalyzing the oxidative ring opening of 3-hydroxyanthranilate to 2-amino-3-carboxymuconate semialdehyde. HAAO may play a role in disorders that are associated with varying levels of QUIN expression, and is widelyMore Product Information