Anti-HAAO Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-11695R-CY5
BOSSBS-11695R-CY5EA 10550 NOK
BOSSBS-11695R-CY5
Anti-HAAO Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
The 3-HAO family of proteins consists of intramolecular dioxygenases that contain non-heme ferrous iron. HAAO (3-hydroxyanthranilate 3,4-dioxygenase), also known as HAO or 3-HAO, is a 286 amino acid monomeric cytoplasmic protein that belongs to the 3-HAO family. Utilizing Fe2+ ions as a cofactor, HAAO participates in the formation of quinolinic acid (QUIN), and excitotoxin, by catalyzing the oxidative ring opening of 3-hydroxyanthranilate to 2-amino-3-carboxymuconate semialdehyde. HAAO may play a role in disorders that are associated with varying levels of QUIN expression, and is widelyMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HAAO
Antigen-navn 3-hydroxyanthranilate 3,4-dioxygenase
Environmentally Preferable
Antigen synonymer 3-HAO|HAO
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR