Anti-CALY Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Leverandør: Bioss
BS-11719R-A555
BOSSBS-11719R-A555EA 10450 NOK
BOSSBS-11719R-A555
Anti-CALY Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
Calcyon is a single transmembrane protein that interacts with D1 dopamine receptors. Dopamine is a neurotransmitter that regulates synaptic transmission involved in learning and memory. D1 receptors, the most abundant dopamine receptor in the central nervous system, appear to modulate the activity of D2 dopamine receptors, mediate various behavioural responses, and regulate neuron growth and differentiation. Calcyon is present in neuronal cell bodies and processes of the cortex and hippocampus, and it is especially abundant in pyramidal neurons. Interaction of Calcyon with D1 receptorsMore Product Information