Anti-CSDC2/PIPPIN Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-11823R-A680
BOSSBS-11823R-A680EA 10425 NOK
BOSSBS-11823R-A680
Anti-CSDC2/PIPPIN Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
PIPPIN is a Y-box protein (also called cold-shock (CSD) domain-containing protein) and belongs to a family of highly conserved RNA-binding transcriptional regulators. Predominantly expressed in brain cells and localizing to the nucleus and the cytoplasm, PIPPIN contains two RNA-binding motifs, namely PIP1 and PIP2, and one CSD domain. PIPPIN functions as a nucleic acid binding regulatory factor and is believed to participate in brain maturation. More specifically, PIPPIN binds to the 3 UTR ends of the mRNAs encoding Histone H1 and Histone H3.3. This interaction requires all of the PIPPINMore Product Information