Anti-CSDC2 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-11823R-CY5
BOSSBS-11823R-CY5EA 10550 NOK
BOSSBS-11823R-CY5
Anti-CSDC2 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
PIPPIN is a Y-box protein (also called cold-shock (CSD) domain-containing protein) and belongs to a family of highly conserved RNA-binding transcriptional regulators. Predominantly expressed in brain cells and localizing to the nucleus and the cytoplasm, PIPPIN contains two RNA-binding motifs, namely PIP1 and PIP2, and one CSD domain. PIPPIN functions as a nucleic acid binding regulatory factor and is believed to participate in brain maturation. More specifically, PIPPIN binds to the 3’-UTR ends of the mRNAs encoding Histone H1 and Histone H3.3. This interaction requires all of the PIPPINMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CSDC2
Environmentally Preferable
Antigen-navn Cold Shock Domain Containing C2
Antigen synonymer RNA binding protein PIPPin|Csdc2|Cold shock domain containing C2 RNA binding|CSDC2_HUMAN|Cold shock domain-containing protein C2|RNA-binding protein PIPPin.|Cold shock domain containing protein C2|dJ347H13.2|PIPPIN|CSDC 2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR