Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-11825R-A555
BOSSBS-11825R-A555EA 10250 NOK
BOSSBS-11825R-A555
Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
DOCK 7 is a 2,140 amino acid protein that localizes to developing axons and contains one DHR-1 domain and one DHR-2 domain. Expressed in a variety of tissues, DOCK 7 functions as a guanine nucleotide exchange factor (GEF) that specifically activates Rac 1 and Rac 3 by catalyzing the exchange of bound GDP for free GTP. Multiple isoforms of DOCK 7 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol DOCK7
Environmentally Preferable
Antigen-navn dedicator of cytokinesis 7
Antigen synonymer ZIR2|EIEE23
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 555
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR