Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-11825R-A680
BOSSBS-11825R-A680EA 10250 NOK
BOSSBS-11825R-A680
Anti-DOCK7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
DOCK 7 is a 2140 amino acid protein that localizes to developing axons and contains one DHR-1 domain and one DHR-2 domain. Expressed in a variety of tissues, DOCK 7 functions as a guanine nucleotide exchange factor (GEF) that specifically activates Rac 1 and Rac 3 by catalyzing the exchange of bound GDP for free GTP. Multiple isoforms of DOCK 7 exist due to alternative splicing events.

Type: Primary
Antigen: DOCK7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1401-1500/2140
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn DOCK7
Antigen synonymer mKIAA1771|MGC189434|KIAA1771 |Dedicator of cytokinesis protein 7|ZIR2|dedicator of cytokinesis 7|Gm430|RP23 329P19.2|DOCK7_HUMAN
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet Human,Rat
Vert Rabbit
Gen-ID 85440
Isotype IgG
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 1401-1500/2140
Immunogen KLH conjugated synthetic peptide derived from human DOCK7
Skjema Liquid
Molekylvekt 242 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR