Anti-LDB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Bioss
BS-11868R-HRP
BOSSBS-11868R-HRPEA 10550 NOK
BOSSBS-11868R-HRP
Anti-LDB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The LIM-only (LMO) proteins, LMO1 and LMO2, are nuclear factors that are characterized by a conserved LIM domain. The LIM domain consists of a cysteine-rich zinc-binding motif that is present in a variety of transcription factors, including the LIM homeobox (LHX) proteins expressed in the central nervous system and involved in cell differentiation. LMO1 and LMO2 are expressed in the adult CNS in a cell type-specific manner, where they are differentially regulated by neuronal activity and are involved in regulating the cellular differentiated phenotype of neurons. LMO2 lacks a specific DNA-More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol LDB1
Environmentally Preferable
Antigen-navn LIM Domain binding 1
Antigen synonymer LIM domain-binding protein 1|CLIM 2|LIM Domain Binding 1|NLI|LIM domain binding factor CLIM2|Carboxyl-terminal LIM domain-binding protein 2|ldb1|Carboxyl Terminal LIM Domain Binding 2|CLIM-2|LDB-1|LIM domain-binding factor CLIM2|hLdb1|LDB1_HUMAN|Nuclear LIM Domain Interactor|Nuclear LIM interactorxldb1.
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR