Anti-MTPN Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Leverandør: Bioss
BS-11891R-A350
BOSSBS-11891R-A350EA 10450 NOK
BOSSBS-11891R-A350
Anti-MTPN Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Myotrophin (V-1 protein) is a ubiquitously expressed cytoplasmic protein that can translocate to the nucleus during sustained NFkB activation. The gene encoding for this protein localizes to chromosome 7q33. Myotrophin may be involved in cerebellar morphogenesis and contains an acetylated N-terminus and 2.5 internal 33 amino acid ankyrin repeats. It is important in the differentiation of cerebellar neurons, particularly of granule cells. The 117 amino acid protein has been associated with, and able to induce, cardiac hypertrophy. Myotrophin increases protooncogene, ANF and Beta-Myosin heavyMore Product Information