Anti-MTPN Rabbit Polyclonal Antibody (Cy3®)
Leverandør: Bioss
BS-11891R-CY3
BOSSBS-11891R-CY3EA 10550 NOK
BOSSBS-11891R-CY3
Anti-MTPN Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Myotrophin (V-1 protein) is a ubiquitously expressed cytoplasmic protein that can translocate to the nucleus during sustained NFkB activation. The gene encoding for this protein localizes to chromosome 7q33. Myotrophin may be involved in cerebellar morphogenesis and contains an acetylated N-terminus and 2.5 internal 33 amino acid ankyrin repeats. It is important in the differentiation of cerebellar neurons, particularly of granule cells. The 117 amino acid protein has been associated with, and able to induce, cardiac hypertrophy. Myotrophin increases protooncogene, ANF and Beta-Myosin heavyMore Product Information