Anti-RAMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-11971R-A680
BOSSBS-11971R-A680EA 10250 NOK
BOSSBS-11971R-A680
Anti-RAMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

Type: Primary
Antigen: RAMP2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-175/175
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn RAMP2
Antigen synonymer RAMP2|Receptor activity-modifying protein 2|Calcitonin-receptor-like receptor activity-modifying protein 2|CRLR activity-modifying protein 2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet
Vert Rabbit
Gen-ID 10266
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 81-175/175
Immunogen KLH conjugated synthetic peptide derived from human RAMP2
Skjema Liquid
Molekylvekt 15 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR