Anti-RAMP2 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-11971R-CY5
BOSSBS-11971R-CY5EA 10350 NOK
BOSSBS-11971R-CY5
Anti-RAMP2 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAMP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR