Anti-RAMP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Bioss
BS-11971R-HRP
BOSSBS-11971R-HRPEA 10350 NOK
BOSSBS-11971R-HRP
Anti-RAMP2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RAMP2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol RAMP2
Environmentally Preferable
Antigen-navn Receptor activity-modifying protein 2
Antigen synonymer RAMP2|Receptor activity-modifying protein 2|Calcitonin-receptor-like receptor activity-modifying protein 2|CRLR activity-modifying protein 2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 10266
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunogen 90-145/175
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR