Anti-RAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-11972R-A488
BOSSBS-11972R-A488EA 10250 NOK
BOSSBS-11972R-A488
Anti-RAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Plays a role in cardioprotection by reducing cardiac hypertrophy and perivascular fibrosis in a GPER1-dependent manner. Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) and GPER1 to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR