Anti-RAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-11972R-A680
BOSSBS-11972R-A680EA 10250 NOK
BOSSBS-11972R-A680
Anti-RAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Plays a role in cardioprotection by reducing cardiac hypertrophy and perivascular fibrosis in a GPER1-dependent manner. Transports the calcitonin gene-related peptide type 1 receptor (CALCRL) and GPER1 to the plasma membrane. Acts as a receptor for adrenomedullin (AM) together with CALCRL.

Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-148/148
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR