Anti-CNG4/GARP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-12093R-A488
BOSSBS-12093R-A488EA 10450 NOK
BOSSBS-12093R-A488
Anti-CNG4/GARP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Cyclic nucleotide-gated (CNG) cation channels are heteromeric complexes made up of principal alpha and modulatory beta subunits (1,2). The alpha subunits consist of CNG1-3 and form functional cation channels by themselves (1,2). The beta subunits consist of CNG4-6 and, unlike the alpha subunits, do not form functional channels, but rather modify the properties of channels (1,2). CNG channels are essential components of olfactory and visual transduction (1,2). In olfactory neurons, CNG2, CNG4.3 and CNG5 form Ca2+ permeable channels, which open and depolarize the cell in response to cAMP (1-3).More Product Information