Anti-CNG4/GARP Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-12093R-CY5
BOSSBS-12093R-CY5EA 10550 NOK
BOSSBS-12093R-CY5
Anti-CNG4/GARP Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Cyclic nucleotide-gated (CNG) cation channels are heteromeric complexes made up of principal alpha and modulatory beta subunits (1,2). The alpha subunits consist of CNG1-3 and form functional cation channels by themselves (1,2). The beta subunits consist of CNG4-6 and, unlike the alpha subunits, do not form functional channels, but rather modify the properties of channels (1,2). CNG channels are essential components of olfactory and visual transduction (1,2). In olfactory neurons, CNG2, CNG4.3 and CNG5 form Ca2+ permeable channels, which open and depolarize the cell in response to cAMP (1-3).More Product Information